Kaliumnitriet 96+% ACS

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

423060050 423065000 423060025
ACRO423060050EA 20.8 EUR
ACRO423060050 ACRO423065000 ACRO423060025
Kaliumnitriet 96+% ACS
Kaliumnitriet
Formule: KNO₂
Smeltpunt: 387 °C
Dichtheid: 1,92 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011408
CAS nummer: 7758-09-0
EINECS: 231-832-4
UN: 1488
ADR: 5.1,II
Merck Index: 13,07734

Order Now

Specification Test Results

Appearance White to light yellow Powder or crystals
Titration with KMnO4 ≥96.0 %
Chloride (Cl) ≤300 ppm
Sulfate (SO4) ≤100 ppm
Calcium (Ca) ≤50 ppm
Iron (Fe) ≤10 ppm
Magnesium (Mg) ≤20 ppm
Sodium (Na) ≤5000 ppm
Heavy metals (as Pb) ≤10 ppm
Insoluble matter ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR