Aluminiumkaliumsulfaat dodecahydraat 98+% ACS

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

423260050 423265000 423260025
ACRO423260050EA 22.3 EUR
ACRO423260050 ACRO423265000 ACRO423260025
Aluminiumkaliumsulfaat dodecahydraat 98+% ACS
Aluminium potassium sulphate dodecahydrate
Formule: AlK(SO₄)₂·12H₂O
Smeltpunt: 92 °C
Dichtheid: 1,72 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149143
CAS nummer: 7784-24-9
EINECS: 233-141-3
Merck Index: 12,00373

Order Now

Specification Test Results

Appearance White Crystals
Titration Complexometric 98.0 to 102.0 %
Chloride (Cl) ≤5 ppm
Ammonium (NH4) ≤50 ppm
Iron (Fe) ≤10 ppm
Sodium (Na) ≤200 ppm
Heavy metals (as Pb) ≤10 ppm
Insoluble matter ≤0.005 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR