Cadmium(II) chloride hemipentahydrate ACS

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

423505000 423500025 423501000
ACRO423505000EA 242 EUR
ACRO423505000 ACRO423500025 ACRO423501000
Cadmium(II) chloride hemipentahydrate ACS
Cadmium(II) chloride hemipentahydrate
Formule: CdCl₂·2.5H₂O
Smeltpunt: 34 °C
Dichtheid: 2,84 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00799071
CAS nummer: 7790-78-5
EINECS: 233-296-7
UN: 2570
ADR: 6.1,III
Merck Index: 13,01617

Order Now

Specification Test Results

Appearance White Crystals and/or chunks
Titration Complexometric 79.5 to 81.0 % (CdCl2)
Nitrate and Nitrite (as NO3) ≤0.003 %
Sulfate (SO4) ≤0.005 %
Ammonium (NH4) ≤0.005 %
Calcium (Ca) ≤0.005 %
Copper (Cu) ≤5 ppm
Iron (Fe) ≤5 ppm
Lead (Pb) ≤0.005 %
Potassium (K) ≤0.02 %
Sodium (Na) ≤0.05 %
Zinc (Zn) ≤0.05 %
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR