Natriumacetaat, anhydrous 99+% ACS

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Acetic acid sodium salt

424255000 424250025
ACRO424255000EA 53.7 EUR
ACRO424255000 ACRO424250025
Natriumacetaat, anhydrous 99+% ACS
Natriumacetaat
Formule: H₃CCOONa
MW: 82.03 g/mol
Kookpunt: 881.4
Smeltpunt: 324 °C
Vlampunt: >250 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00012459
CAS nummer: 127-09-3
EINECS: 204-823-8
Bereik: 01-2119485123-42
Merck Index: 12,08711

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤1.0 % (120°C)
pH 7.0 to 9.2 (5 % at 25.0°C)
Titration with HClO4 ≥99.0 %
Chloride (Cl) ≤0.002 %
Phosphate (PO4) ≤0.001 %
Sulfate (SO4) ≤0.003 %
Calcium (Ca) ≤0.005 %
Iron (Fe) ≤0.001 %
Magnesium (Mg) ≤0.002 %
Heavy metals (ICP-OES) ≤0.001 %
Insoluble matter ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR