Natriumacetaat trihydraat 99+% ACS

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Acetic acid sodium salt trihydrate

424260010 424260250 424260025
ACRO424260010EA 51.5 EUR
ACRO424260010 ACRO424260250 ACRO424260025
Natriumacetaat trihydraat 99+% ACS
Natriumacetaat trihydraat
Formule: H₃CCOONa·3H₂O
Kookpunt: 120 - 123 °C (1013 hPa)
Smeltpunt: 58 °C
Dichtheid: 1,4 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00071557
CAS nummer: 6131-90-4
EINECS: 204-823-8
Merck Index: 13,08642
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=313c1d45673fab192565649f8cbb7ddff555de39&vid=1ad37a966605626c95117735c83b16eaea381fbc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Transparent white Crystals
Infrared spectrum Conforms
pH 7.5 to 9.2 (5 % at 25.0°C)
Titration with HClO4 99.0 to 101.0 %
Chloride (Cl) ≤0.001 %
Phosphate (PO4) ≤5 ppm
Sulfate (SO4) ≤0.002 %
Calcium (Ca) ≤0.005 %
Iron (Fe) ≤5 ppm
Magnesium (Mg) ≤0.002 %
Potassium (K) ≤0.005 %
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.005 %
KMnO4-reducing substance Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR