Sodium tetraphenylborate ACS

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

424450250 424451000 424455000
ACRO424450250EA 161 EUR
ACRO424450250 ACRO424451000 ACRO424455000
Sodium tetraphenylborate ACS
Sodium tetraphenylborate
Formule: (C₆H₅)₄BNa
Smeltpunt: >300 °C
Dichtheid: 1,15 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011494
CAS nummer: 143-66-8
EINECS: 205-605-5
UN: 2811
ADR: 6.1,III
Merck Index: 13,08765

Order Now

Specification Test Results

Appearance White to light beige Powder or crystals
Infrared spectrum Conforms
Loss on drying ≤0.5 % (105°C)
Gravimetry ≥99.5 %
Clarity of solution Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR