N-Boc-DL-valine 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

425780100 425780500
ACRO425780100EA 69.1 EUR
ACRO425780100 ACRO425780500
N-Boc-DL-valine 98%
N-Boc-DL-valine
Formule: C₁₀H₁₉NO₄
Smeltpunt: 112…114 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00069973
CAS nummer: 54895-12-4

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %
NMR consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR