Methyl undecanoate 97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Undecanoic acid methyl ester, Undecylic acid methyl ester

426580050 426580250
ACRO426580050EA 45.7 EUR
ACRO426580050 ACRO426580250
Methyl undecanoate 97%
Methyl undecanoate
Formule: C₁₂H₂₄O₂
MW: 200.32 g/mol
Kookpunt: 246…248 °C (760 mmHg)
Dichtheid: 0.872 g/cm³
Vlampunt: 109 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008957
CAS nummer: 1731-86-8

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR