2-(3-Aminophenyl)acetic acid 97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

427610010 427610100
ACRO427610010EA 39.2 EUR
ACRO427610010 ACRO427610100
2-(3-Aminophenyl)acetic acid 97%
2-(3-Aminophenyl)acetic acid

Appearance: Off-white to light yellow Crystalline powder

Formule: C₈H₉NO₂
Smeltpunt: 147…150 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00075058
CAS nummer: 14338-36-4

Order Now

Specification Test Results

Appearance Off-white to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 147 to 154 °C
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR