Glycine benzyl ester hydrochloride (H-Gly-OBzl.HCl) 97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

428540010 428540050 428540250
ACRO428540010EA 30.2 EUR
ACRO428540010 ACRO428540050 ACRO428540250
Glycine benzyl ester hydrochloride (H-Gly-OBzl.HCl) 97%
Glycine benzyl ester hydrochloride (H-Gly-OBzl.HCl)
Formule: C₉H₁₂ClNO₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00035442
CAS nummer: 2462-31-9

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
HPLC ≥96.0 %
NMR consistent with structure
Iron (Fe) ≤0.0001 %
Heavy metals (as Pb) ≤1 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR