N-Methoxy-N-methylbenzamide 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

436560050
ACRO436560050EA 73.4 EUR
ACRO436560050
N-Methoxy-N-methylbenzamide 98%
N-Methoxy-N-methylbenzamide
Formule: C₉H₁₁NO₂
MW: 165.19 g/mol
Kookpunt: 85 °C (1 mmHg)
Vlampunt: 113 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00075320
CAS nummer: 6919-61-5

Order Now

Specification Test Results

Appearance Colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR