Gold trichloride 99.99+% (trace metals basis)
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
Waarschuwing
Synoniemen: Gold(III) chloride, Gold (III) chloride
437080010 437080050
ACRO437080010EA 284 EUR
ACRO437080010 ACRO437080050
Gold trichloride 99.99+% (trace metals basis)
Gold trichloride
Formule: AuCl₃
MW: 303.32 g/mol
Smeltpunt: 254 °C
Dichtheid: 4.3
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00014172
CAS nummer: 13453-07-1
Merck Index: 13,04536
Order Now
Specification Test Results
Appearance Red to brown Crystalline powder or crystals
Assay 63.0 to 66.9 % (Au)
Total trace metal impurities ≤100 ppm