1-Boc-4-(2-chloroethyl)piperazine 97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

438240010 438240050
ACRO438240010EA 106 EUR
ACRO438240010 ACRO438240050
1-Boc-4-(2-chloroethyl)piperazine 97%
1-Boc-4-(2-chloroethyl)piperazine
Formule: C₁₁H₂₁ClN₂O₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD08443993
CAS nummer: 208167-83-3

Order Now

Specification Test Results

Appearance White to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 62 to 68 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR