FMOC-O-tert-butyl-L-tyrosine 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

439090250 439090100
ACRO439090250EA 0 EUR
ACRO439090250 ACRO439090100
FMOC-O-tert-butyl-L-tyrosine 98%
FMOC-O-tert-butyl-L-tyrosine
Formule: C₂₈H₂₉NO₅
Smeltpunt: 150…156 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00037129
CAS nummer: 71989-38-3

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Specific optical rotation -27° to -31° (20°C, 589 nm) (c=1 in DMF)

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR