(S)-2-((5-Fluoro-2,4-dinitrophenyl)amino)propanamide 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

439341000
ACRO439341000EA 249 EUR
ACRO439341000
(S)-2-((5-Fluoro-2,4-dinitrophenyl)amino)propanamide 98%
(S)-2-((5-Fluoro-2,4-dinitrophenyl)amino)propanamide
Formule: C₉H₉FN₄O₅
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00042049
CAS nummer: 95713-52-3

Order Now

Specification Test Results

Appearance Yellow to orange Powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Specific optical rotation +54.0° to +58.0° (20°C, 589 nm) (c=1, acetone)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR