N-Methylisopropylamine 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

439650100 439650500
ACRO439650100EA 54.9 EUR
ACRO439650100 ACRO439650500
N-Methylisopropylamine 98%
N-Methylisopropylamine
Formule: C₄H₁₁N
Kookpunt: 50…53 °C (760 mmHg)
Dichtheid: 0,702 g/cm³
Vlampunt: –32 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00042859
CAS nummer: 4747-21-1
UN: 2734
ADR: 8,I

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3830 to 1.3850 (20°C, 589 nm)
Titration with HClO4 ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR