1-Bromo-2-naphthoic acid 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

440390010
ACRO440390010EA 151 EUR
ACRO440390010
1-Bromo-2-naphthoic acid 98%
1-Bromo-2-naphthoic acid
Formule: C₁₁H₇BrO₂
Smeltpunt: 190…192 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00021408
CAS nummer: 20717-79-7

Order Now

Specification Test Results

Appearance White to yellow to light brown Powder
Infrared spectrum Conforms
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR