Nα-Fmoc-N-δ-trityl-L-glutamine 95%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

440640050 440640250
ACRO440640050EA 0 EUR
ACRO440640050 ACRO440640250
Nα-Fmoc-N-δ-trityl-L-glutamine 95%
N-alpha-Fmoc-N-delta-trityl-L-glutamine
Formule: C₃₉H₃₄N₂O₅
Smeltpunt: 131…144 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00077056
CAS nummer: 132327-80-1

Order Now

Specification Test Results

Appearance White to yellow Powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Specific optical rotation -12.5° to -15.5° (20°C, 589 nm) (c=1 in DMF)

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR