Palladium(II) acetate 99.9% (trace metals basis)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Palladium diacetate, Palladium (II) acetate

441390010 441390050
ACRO441390010EA 155 EUR
ACRO441390010 ACRO441390050
Palladium(II) acetate 99.9% (trace metals basis)
Palladium(II) acetate
Formule: C₄H₆O₄Pd
MW: 224.51 g/mol
Smeltpunt: 205 °C (ontbinding)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00012453
CAS nummer: 3375-31-3

Order Now

Specification Test Results

Appearance Orange to brown Powder
Assay 46.9 to 48.5 % (Pd)
Total trace metal impurities ≤1000 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR