Petroleum spirit, 100…140 °C, Technical Grade

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Gasoline, Petroleum ether, Petrol, Benzine

444360250
ACRO444360250EA 153 EUR
ACRO444360250
Petroleum spirit, 100…140 °C, Technical Grade
Petroleum spirit
Kookpunt: 100…140 °C (760 mmHg)
Smeltpunt: –30 °C
Dichtheid: 0,74 g/cm³
Vlampunt: 2 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00081849
CAS nummer: 64742-49-0
UN: 3295
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Refractive index 1.4070 to 1.4150 (20 °C, 589 nm)
Boiling point 98 to 140 °C (760 mmHg)
Density 0.737 to 0.747 kg/l


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR