4-(Tetrazol-5-yl)phenylboronic acid 97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

445570010 445570050
ACRO445570010EA 120 EUR
ACRO445570010 ACRO445570050
4-(Tetrazol-5-yl)phenylboronic acid 97%
4-(Tetrazol-5-yl)phenylboronic acid
Formule: C₇H₇BN₄O₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD06739099
CAS nummer: 179942-55-3

Order Now

Specification Test Results

Appearance White to off-white Solid
Infrared spectrum Conforms
HPLC ≥95.0 %
NMR (1H-NMR) Consistent with structure

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR