1,2-Dichloorbenzeen 99+% for HPLC

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

445640010 445641000
ACRO445640010EA 102 EUR
ACRO445640010 ACRO445641000
1,2-Dichloorbenzeen 99+% for HPLC
1,2-Dichloorbenzeen
Formule: C₆H₄Cl₂
Kookpunt: 180,5 °C (1013 hPa)
Smeltpunt: –15 °C
Dichtheid: 1,3 g/cm³
Vlampunt: 67 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000535
CAS nummer: 95-50-1
UN: 1591
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear liquid
Water ≤0.1 %
GC ≥99.0 %
UV at 380 nm A: ≤0.01
Color scale ≤10 APHA
Residue after evaporation ≤0.0005 %
Acidity ≤0.0003 meq/g
Alkalinity ≤0.0005 meq/g

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR