Piperidine-2,4-dione 97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: 2,4-Piperidinedione, 2,4-Dioxopiperidine

446730010 446730050
ACRO446730010EA 144 EUR
ACRO446730010 ACRO446730050
Piperidine-2,4-dione 97%
Piperidine-2,4-dione
Formule: C₅H₇NO₂
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD08704814
CAS nummer: 50607-30-2

Order Now

Specification Test Results

Appearance White to yellow to light brown Powder or solid
Infrared spectrum Conforms
Melting point 98 to 104 °C
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR