Magnesium chloride hexahydrate 99% voor de biochemie

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

447150025 447155000
ACRO447150025EA 206 EUR
ACRO447150025 ACRO447155000
Magnesium chloride hexahydrate 99% voor de biochemie
Magnesium chloride hexahydrate
Formule: MgCl₂·6H₂O
Kookpunt: 1412 °C (1013 hPa)
Smeltpunt: 117 °C
Dichtheid: 1,57 g/cm³ (25 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149781
CAS nummer: 7791-18-6

Order Now

Specification Test Results

Appearance White Crystalline powder
Titration Complexometric ≥98.5 %
UV (1 M in water)
pH 5.0 to 6.5
Phosphate (PO4) ≤0.0005 %
Sulfate (SO4) ≤0.002 %
Aluminium (Al) ≤0.0002 %
Iron (Fe) ≤0.0005 %
Lead (Pb) ≤0.0005 %
Zinc (Zn) ≤0.001 %
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.005 % (in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR