(±)-1,2-Propanediol ACS

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

447410025 447411000 447415000
ACRO447410025EA 179 EUR
ACRO447410025 ACRO447411000 ACRO447415000
(±)-1,2-Propanediol ACS
(±)-1,2-Propanediol
Formule: CH₃CH(OH)CH₂OH
Kookpunt: 187 °C (760 mmHg)
Smeltpunt: –60 °C
Dichtheid: 1,03 g/cm³
Vlampunt: 99 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00064272
CAS nummer: 57-55-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f2af195b87767feb8d1baca34925153d6f97ae2d&vid=a598a9042847dc3ce52daed1a9326b592b163170&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear viscous liquid
Water ≤0.2 %
GC ≥99.5 %
Color scale ≤10 APHA
Residue after ignition ≤0.005 %
Chloride (Cl) ≤1 ppm
Titrable acid ≤0.0005 meq/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR