Potassium dihydrogen phosphate 99% for analysis

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Monopotassiumphosphate, prim.-Potassium phosphate, Monopotassium phosphate, Potassium phosphate monobasic, Monobasic Potassium Phosphate, Potassium dihydrogen orthophosphate

447670010 447670050
ACRO447670010EA 90.5 EUR
ACRO447670010 ACRO447670050
Potassium dihydrogen phosphate 99% for analysis
Potassium dihydrogen phosphate
Formule: KH₂PO₄
Kookpunt: ~ 100 °C (1013 hPa)
Smeltpunt: 253 °C
Dichtheid: 2,3 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011401
CAS nummer: 7778-77-0

Order Now

Specification Test Results

Appearance White Crystalline powder
Loss on drying ≤0.2 %
pH 4.2 to 4.5 (5 % solution)
Titration with NaOH ≥98.5 %
Chloride (Cl) ≤0.001 %
Sulfate (SO4) ≤0.003 %
Heavy metals (as Pb) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR