2-Quinolinecarbonitrile 97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

449580010 449580050
ACRO449580010EA 55.8 EUR
ACRO449580010 ACRO449580050
2-Quinolinecarbonitrile 97%
2-Quinolinecarbonitrile
Formule: C₁₀H₆N₂
Kookpunt: 324 °C (1013 hPa)
Smeltpunt: 93…95 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00134341
CAS nummer: 1436-43-7
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to yellow to beige Powder
Infrared spectrum Conforms
Melting point 92 to 96 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR