4,4'-Methylenebis(2-methylcyclohexylamine) (mixture of isomers) mixture of isomers 99% (mixture of isomers)

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

450240010 450242500
ACRO450240010EA 256 EUR
ACRO450240010 ACRO450242500
4,4'-Methylenebis(2-methylcyclohexylamine) (mixture of isomers) mixture of isomers 99% (mixture of isomers)
4,4'-Methylenebis(2-methylcyclohexylamine) (mixture of isomers)
Formule: C₁₅H₃₀N₂
Kookpunt: 93…100 °C (760 mmHg)
Dichtheid: 0,94 g/cm³
Vlampunt: 113 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00075535
CAS nummer: 6864-37-5
UN: 2927
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4980 to 1.5000 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR