Kanamycin sulphate

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

450810100 450810500 450811000
ACRO450810100EA 215 EUR
ACRO450810100 ACRO450810500 ACRO450811000
Kanamycin sulphate
Kanamycin sulphate
Formule: C₁₈H₃₈N₄O₁₅S
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00070253
CAS nummer: 25389-94-0

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Assay ≥760 µg/mg (on dried basis)
Loss on drying ≤4.0 %
Solubility Clear colorless
TLC Complies
Residue after ignition ≤1.0 %
pH 6.5 to 8.5 (1 % in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR