Ethanolamine ACS

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

451760010 451760025 451762500
ACRO451760010EA 67.1 EUR
ACRO451760010 ACRO451760025 ACRO451762500
Ethanolamine ACS
Ethanolamine
Formule: NH₂CH₂CH₂OH
Kookpunt: 170 °C (760 mmHg)
Dichtheid: 1,012 g/cm³
Vlampunt: 92 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008183
CAS nummer: 141-43-5
UN: 2491
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=82bfb6ea7cf5dde341c60ff9cf752bc5c422eb28&vid=b387e8c39029da4ea050ba6dcfb6123d81ab03c8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Water ≤0.30 %
GC ≥99.0 %
Color scale ≤15 APHA
Iron (Fe) ≤5 ppm
Heavy metals (as Pb) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR