Tin(IV) oxide 99%, pure

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

451810025 451811000 451810010
ACRO451810025EA 270 EUR
ACRO451810025 ACRO451811000 ACRO451810010
Tin(IV) oxide 99%, pure
Tin(IV) oxide
Formule: O₂Sn
Dichtheid: 6,95 g/cm³
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011244
CAS nummer: 18282-10-5

Order Now

Specification Test Results

Appearance White to light grey Powder
Purity ≥98.5 % (metals basis)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR