3-(Trifluoromethyl)piperidine 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

454820010
ACRO454820010EA 123 EUR
ACRO454820010
3-(Trifluoromethyl)piperidine 98%
3-(Trifluoromethyl)piperidine
Formule: C₆H₁₀F₃N
Kookpunt: 128…130 °C
Vlampunt: 28 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD02183561
CAS nummer: 768-31-0
UN: 1992
ADR: 3,III

Order Now

Specification Test Results

Appearance Colorless to light brown Liquid
Infrared spectrum Conforms
GC ≥97.5 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR