Amphotericin B 85%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

455490010 455490050
ACRO455490010EA 128 EUR
ACRO455490010 ACRO455490050
Amphotericin B 85%
Amphotericin B
Formule: C₄₇H₇₃NO₁₇
Smeltpunt: >170 °C
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00877763
CAS nummer: 1397-89-3

Order Now

Specification Test Results

Appearance Yellow to orange Powder
UV spectrum Complies
Water ≤10 %
HPLC ≥80 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR