Rifampicin 95%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

455620010 455620050 455620250
ACRO455620010EA 35.3 EUR
ACRO455620010 ACRO455620050 ACRO455620250
Rifampicin 95%
Rifampicin
Formule: C₄₃H₅₈N₄O₁₂
Smeltpunt: 183 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00151389
CAS nummer: 13292-46-1

Order Now

Specification Test Results

Appearance Light orange to red-brown Powder
Infrared spectrum Conforms
Loss on drying ≤2 % (100 mg, 60°C, 4 h) (vacuum)
HPLC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR