2-(Isocyanatomethyl)furan 97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Furfuryl isocyanate

456490010 456490050
ACRO456490010EA 58.7 EUR
ACRO456490010 ACRO456490050
2-(Isocyanatomethyl)furan 97%
2-(Isocyanatomethyl)furan
Formule: C₆H₅NO₂
MW: 123.11 g/mol
Smeltpunt: 157…158 °C
Dichtheid: 1.148 g/cm³
Vlampunt: 54.5 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD03180268
CAS nummer: 71189-15-6
UN: 1993
ADR: 3 III

Order Now

Specification Test Results

Appearance Clear colorless to brown Liquid
Infrared spectrum Conforms
Refractive index 1.4750 to 1.4770 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR