Fluvastatin sodium salt

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: XU 62-320, rel-(3S,5R,E)-7-(3-(4-Fluorophenyl)-1-isopropyl-1H-indol-2-yl)-3,5-dihydroxyhept-6-enoic acid sodium salt, Sodium rel-(3R,5S,E)-7-(3-(4-fluorophenyl)-1-isopropyl-1H-indol-2-yl)-3,5-dihydroxyhept-6-enoate

458010010 458010050
ACRO458010010EA 79.4 EUR
ACRO458010010 ACRO458010050
Fluvastatin sodium salt
Fluvastatin sodium salt
Formule: C24 H25FNNaO4
MW: 433.46 g/mol
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00929076
CAS nummer: 93957-55-2

Order Now

Specification Test Results

Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Water ≤4 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR