Econazole 99%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

458630050 458630250
ACRO458630050EA 50.4 EUR
ACRO458630050 ACRO458630250
Econazole 99%
Econazole
Formule: C₁₈H₁₅Cl₃N₂O
Smeltpunt: 91…94 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00800993
CAS nummer: 27220-47-9

Order Now

Specification Test Results

Appearance White to off-white powder Powder
Infrared spectrum Conforms
Melting point 88 to 92 °C
Loss on drying ≤0.5 % (1 g, 60 °C, 4 h)
HPLC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR