L-Lysine methyl ester dihydrochloride 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: H-Lys-OMe·2HCl, H-Lys-OMe.2HCl

459270050 459270250 459271000
ACRO459270050EA 27.4 EUR
ACRO459270050 ACRO459270250 ACRO459271000
L-Lysine methyl ester dihydrochloride 98%
L-Lysine methyl ester dihydrochloride
Formule: C₇H₁₆N₂O₂
MW: 233.13 g/mol
Smeltpunt: 200 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00039067
CAS nummer: 26348-70-9

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Specific optical rotation +21° to +25° (20°C, 589 nm) (1% in MeOH)
Titration with HClO4 ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR