Hexyl nicotinate 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

460090050 460090250
ACRO460090050EA 30.4 EUR
ACRO460090050 ACRO460090250
Hexyl nicotinate 98%
Hexyl nicotinate
Formule: C₁₂H₁₇NO₂
Kookpunt: 329 °C (760 mmHg)
Dichtheid: 1,02 g/cm³
Vlampunt: 128 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00023587
CAS nummer: 23597-82-2

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4890 to 1.4910 (20°C, 589 nm)
Water ≤0.3 %
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR