Gatifloxacin 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

460230010 460230050 460230250
ACRO460230010EA 42.9 EUR
ACRO460230010 ACRO460230050 ACRO460230250
Gatifloxacin 98%
Gatifloxacin
Formule: C₁₉H₂₂FN₃O₄
Smeltpunt: 185…187 °C
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00895399
CAS nummer: 112811-59-3

Order Now

Specification Test Results

Appearance White to light yellow Powder
Infrared spectrum Conforms
Loss on drying 4 % to 8 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR