Gliclazide

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

461070010 461070050
ACRO461070010EA 40.8 EUR
ACRO461070010 ACRO461070050
Gliclazide
Gliclazide
Formule: C₁₅H₂₁N₃O₃S
Smeltpunt: 163…169 °C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00409893
CAS nummer: 21187-98-4

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 163 to 169 °C
Loss on drying ≤0.5 % (1 g, 105 °C)
HPLC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR