Pyridostigmine bromide 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

461190010 461190050 461190250
ACRO461190010EA 56.1 EUR
ACRO461190010 ACRO461190050 ACRO461190250
Pyridostigmine bromide 98%
Pyridostigmine bromide
Formule: C₉H₁₃BrN₂O₂
Smeltpunt: 154 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00079283
CAS nummer: 101-26-8
UN: 2811
ADR: 6.1,II

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 153 to 157 °C
Loss on drying ≤2 % (1 g, 105 °C)
Titration with HClO4 97.5 to 102.5 % (on dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR