Prasugrel 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

461480010 461482500
ACRO461480010EA 137 EUR
ACRO461480010 ACRO461482500
Prasugrel 98%
Prasugrel
Formule: C₂₀H₂₀FNO₃S
Smeltpunt: 110…121 °C
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD09954140
CAS nummer: 150322-43-3

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Water ≤0.5 %
HPLC ≥97.5 %
Residue after ignition ≤0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR