Dimethyl (1-diazo-2-oxopropyl)phosphonate 95%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Ohira-Bestmann Reagent, 1-Diazoacetonylphosphonic Acid Dimethyl Ester, Dimethyl 1-Diazoacetonylphosphonate, (1-Diazo-2-oxopropyl)phosphonic Acid Dimethyl Ester

466020010 466020050
ACRO466020010EA 99.8 EUR
ACRO466020010 ACRO466020050
Dimethyl (1-diazo-2-oxopropyl)phosphonate 95%
Dimethyl (1-diazo-2-oxopropyl)phosphonate
Formule: C₅H₉N₂O₄P
MW: 192.11 g/mol
MDL Nummer: MFCD07368360
CAS nummer: 90965-06-3

Order Now

Specification Test Results

Appearance (Color) Clear light yellow to brown
Appearance (Form) Liquid
Refractive index 1.4770 to 1.4820 (20°C, 589 nm)
HPLC >=94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR