Palladium standard solution, 1,000 mg/L Pd in 5% nitric acid standard for ICP
Leverancier: ACTIVE MOTIF
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
C042.5NP.L1 C042.5NP.L25 C042.5NP.L5
ACSDC042.5NP.L1EA 191 EUR
ACSDC042.5NP.L1 ACSDC042.5NP.L25 ACSDC042.5NP.L5
Palladium standard solution, 1,000 mg/L Pd in 5% nitric acid standard for ICP
Standaarden Kationische standaarden Palladium standaarden