Calcium standard solution, 10000 mg/L Ca in 5% nitric acid standard for ICP
Leverancier: CPACHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
C109.5NP.L1 C109.5NP.L25 C109.5NP.L5
ACSDC109.5NP.L1EA 192 EUR
ACSDC109.5NP.L1 ACSDC109.5NP.L25 ACSDC109.5NP.L5
Calcium standard solution, 10000 mg/L Ca in 5% nitric acid standard for ICP
Standaarden Kationische standaarden Calcium standaarden