Cobalt standard solution, 10000 mg/L Co in 5% nitric acid standard for ICP
Leverancier: ACTIVE MOTIF
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
C112.5NP.L1 C112.5NP.L25 C112.5NP.L5
ACSDC112.5NP.L1EA 192 EUR
ACSDC112.5NP.L1 ACSDC112.5NP.L25 ACSDC112.5NP.L5
Cobalt standard solution, 10000 mg/L Co in 5% nitric acid standard for ICP
Standaarden Kationische standaarden Kobalt standaarden