Phosphorus standard solution, 10,000 mg/L P in 0,05% sulphuric acid standard for ICP
Leverancier: CPACHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
C140.05SP C140.005SP.L1 C140.005SP.L25 C140.005SP.L5
ACSDC140.05SPEA 165 EUR
ACSDC140.05SP ACSDC140.005SP.L1 ACSDC140.005SP.L25 ACSDC140.005SP.L5
Phosphorus standard solution, 10,000 mg/L P in 0,05% sulphuric acid standard for ICP
Standaarden Kationische standaarden Fosfor standaarden