Tissue forceps, Aesculap® SUSI®
Leverancier: Aesculap
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BD557SU
AESCBD557SUEA 75.1 EUR
AESCBD557SU
Tissue forceps, Aesculap® SUSI®
Pincetten
These disposable forceps are made from a high performance polymer. The special composition of this high-grade polymer ensures an outstanding quality and provides optimal mechanical properties.
  • Material ensures a safe, easy and environmentally friendly removal through incineration in the regular hospital waste removal system
  • Single use: Reduces the risk of cross-contamination
  • 1×2 teeth
  • Sterile single packed in dispenser box
Order Now
Specificaties
Environmentally Preferable