Suction cannulae, Aesculap®
Leverancier: Aesculap
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
MD124R MD125R MD127R MD129R
AESCMD124REA 0 EUR
AESCMD124R AESCMD125R AESCMD127R AESCMD129R
Suction cannulae, Aesculap®
Canules
Suction cannulae with finger cut-off and stylet.
  • Author: Frazier
  • Thumb piece 90° to curve
  • With LUER hub
Order Now
Specificaties
Environmentally Preferable